accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOONH2H2N

CAS RN 68960-97-4

IUPAC Name: 3,6,9,12-tetraoxatetradecane-1,14-diamine
Synonyms: 3,6,9,12-tetraoxatetradecane-1,14-diamine
Molecular Weight (g/mol): 236.31
Molecular Formula: C10H24N2O4
InChi Key: IFZOPNLVYZYSMQ-UHFFFAOYSA-N
SMILES: NCCOCCOCCOCCOCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results