CAS RN 69-65-8
Thermo Scientific Chemicals D-Mannitol, ACS
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 IUPAC Name: (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Thermo Scientific Chemicals D-Mannitol, 98+%
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 SMILES: OCC(O)C(O)C(O)C(O)CO
D-Mannitol, ACS reagent
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
D-Mannitol (Powder/Certified ACS), Fisher Chemical™
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Thermo Scientific Chemicals D-Mannitol, 97+%
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 IUPAC Name: (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
MilliporeSigma™ Calbiochem™ D(-)Mannitol
For Life Science research application.
Mannitol, Powder, Multi-Compendial, U.S.P., J.T. Baker™
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 SMILES: OCC(O)C(O)C(O)C(O)CO
Mannitol, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 IUPAC Name: (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
D-Mannitol, MP Biomedicals
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Mannitol ACS AR Powder, Macron Fine Chemicals™
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Mannitol, Powder, USP, 97-102%, Spectrum™ Chemical
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
D-Mannitol, MP Biomedicals™
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: Mannite IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
D-mannitol, 99%, MP Biomedicals™
CAS: 69-65-8 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00064287 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-mannitol,mannitol,mannite,osmitrol,manna sugar,osmofundin,cordycepic acid,mannit,mannazucker,mannidex PubChem CID: 6251 ChEBI: CHEBI:16899 SMILES: OCC(O)C(O)C(O)C(O)CO
- 1
- 2