Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Mannitol, MP Biomedicals

Catalog No. MP21022485 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21022485 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21022485 Supplier MP Biomedicals, Inc Supplier No. 02102248.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • D-Mannitol used with boric acid in the manufacturing of dry electrolytic condensers for radio applications
  • In pharmacy as excipient and diluent for solids and liquids
  • In analytical chemistry for boron determinations
  • In the manufuring of mannitol hexanitrate
  • It is used as an anticaking and free-flow agent, flavoring agent, lubricant and release agent, stabilizer and thickener and nutritive sweetener in the food industry
  • Also used as a diuretic

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 163°C to 172°C
Boiling Point 290°C to 295°C (3.5mm)(literature)
Linear Formula C6H14O6
Quantity 500 g
Solubility Information Soluble in water (1 g/5.5 mL), alcohol (1 g/83 mL), glycerol (1 g/18 mL); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.
Formula Weight 182.2
Percent Purity ≥96%
Physical Form Powder
Chemical Name or Material D-MANNITOL
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.