accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 696-07-1

IUPAC Name: 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-iodouracil
Molecular Weight (g/mol): 237.98
Molecular Formula: C4H3IN2O2
InChi Key: KSNXJLQDQOIRIP-UHFFFAOYSA-N
SMILES: IC1=CNC(=O)NC1=O
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)