Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 5-Iodouracil, 99%

Catalog No. AC228160500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228160500 50 g Glass bottle
AC228160100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC228160500 Supplier Thermo Scientific Chemicals Supplier No. 228160500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 696-07-1 | C4H3IN2O2 | 237.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-07-1
Molecular Formula C4H3IN2O2
Molecular Weight (g/mol) 237.98
MDL Number MFCD00006020
InChI Key KSNXJLQDQOIRIP-UHFFFAOYSA-N
Synonym 5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 69672
ChEBI CHEBI:43636
IUPAC Name 5-iodo-1H-pyrimidine-2,4-dione
SMILES IC1=CNC(=O)NC1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 270.0°C to 274.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ammonia water and other alkalines
Formula Weight 237.98
Percent Purity 99%
Physical Form Fluffy Powder
Chemical Name or Material 5-Iodouracil
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse
EINECSNumber 211-788-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.