accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(R)O(R)OH(S)OH(S)OH(R)HO

CAS RN 69984-73-2

IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(nonyloxy)oxane-3,4,5-triol
Synonyms: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(nonyloxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 306.4
Molecular Formula: C15H30O6
InChi Key: QFAPUKLCALRPLH-UXXRCYHCSA-N
SMILES: CCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)