Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonyl beta-D-glucopyranoside, 98%

Catalog No. AAJ6600906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6600906 5 g
AAJ6600903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6600906 Supplier Thermo Scientific Chemicals Supplier No. J6600906
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 69984-73-2 | C15H30O6 | 306.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69984-73-2
Molecular Formula C15H30O6
Molecular Weight (g/mol) 306.399
MDL Number MFCD00063300
InChI Key QFAPUKLCALRPLH-UXXRCYHCSA-N
Synonym nonyl beta-d-glucopyranoside, b-nonylglucoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy tetrahydro-2h-pyran-3,4,5-triol, b-d-glucopyranoside, nonyl, nonyl-beta-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy oxane-3,4,5-triol, n-nonyl-beta-d-glucopyranoside, bng, beta-d-glucopyranoside, nonyl, 4gby
PubChem CID 155448
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol
SMILES CCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 5 g
Beilstein 12168
Formula Weight 306.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nonyl beta-D-glucopyranoside
EINECSNumber 615-042-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.