accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONHNHNNNHNHOSI

CAS RN 702675-74-9

IUPAC Name: N-(3-{[5-iodo-4-({3-[(thiophen-2-yl)formamido]propyl}amino)pyrimidin-2-yl]amino}phenyl)pyrrolidine-1-carboxamide
Synonyms: N-{3-[(5-iodo-4-{[3-(thiophen-2-ylformamido)propyl]amino}pyrimidin-2-yl)amino]phenyl}pyrrolidine-1-carboxamide
Molecular Weight (g/mol): 591.47
Molecular Formula: C23H26IN7O2S
InChi Key: VAVXGGRQQJZYBL-UHFFFAOYSA-N
SMILES: IC1=CN=C(NC2=CC(NC(=O)N3CCCC3)=CC=C2)N=C1NCCCNC(=O)C1=CC=CS1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results