Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BX 795
GREENER_CHOICESDP

Catalog No. 431850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-185-0 50 mg
43-181-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-185-0 Supplier R&D Systems™ Supplier No. 4318/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PDPK1 (PDK1) inhibitor

Inhibitor of 3-phosphoinositide-dependent kinase 1 (PDPK1). Inhibits Akt phosphorylation at Thr308 in PC3 cells; also inhibits anchorage-independent growth of PC3 and MDA-MB-468 cells. Exhibits activity at other kinases, including TANK-binding kinase 1 (TBK1), Aurora B and IκB kinase ε (IKKε). Also enhances lentiviral transduction of natural killer (NK) cells by around 3.8-fold.

Chemical Identifiers

CAS 702675-74-9
Molecular Formula C23H26IN7O2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 591.47
Purity >98%
Molecular Weight (g/mol) 591.47
Chemical Name or Material BX 795
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.