accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOH3CCH3CH3H3C(E)

CAS RN 71441-28-6

IUPAC Name: 4-[(1E)-2-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)prop-1-en-1-yl]benzoic acid
Synonyms: ttnpb arotinoid acid tocris-0761 arotinoic acid arotinoid ro-137410 T3757_sigma
Molecular Weight (g/mol): 348.49
Molecular Formula: C24H28O2
InChi Key: FOIVPCKZDPCJJY-JQIJEIRASA-N
SMILES: C\C(=C/C1=CC=C(C=C1)C(O)=O)C1=CC=C2C(=C1)C(C)(C)CCC2(C)C
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results