Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TTNPB
GREENER_CHOICESDP

Catalog No. 076150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-615-0 50 mg
07-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-615-0 Supplier R&D Systems™ Supplier No. 0761/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Retinoic acid analog; RAR agonist

Extremely potent analog of retinoic acid, selective for the retinoic acid receptor (RAR) subtype. Enhances reprogramming efficiency in chemically induced pluripotent stem cells (CiPSCs).

Chemical Identifiers

CAS 71441-28-6
Molecular Formula C24H28O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 348.48
Purity >98%
Molecular Weight (g/mol) 348.486
Chemical Name or Material TTNPB
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.