accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7149-80-6

CH3NH2ClNOO

CAS RN 7149-80-6

IUPAC Name: 5-chloro-4-methyl-2-nitroaniline
Synonyms: 5-chloro-4-methyl-2-nitroaniline
Molecular Weight (g/mol): 186.6
Molecular Formula: C7H7ClN2O2
InChi Key: BBWHBSDZHSQECL-UHFFFAOYSA-N
SMILES: CC1=CC(=C(N)C=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results