accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 724709-68-6

IUPAC Name: 4-(4-phenylbutoxy)-7H-furo[3,2-g]chromen-7-one
Synonyms: 4-(4-phenylbutoxy)furo[3,2-g]chromen-7-one
Molecular Weight (g/mol): 334.37
Molecular Formula: C21H18O4
InChi Key: JJAWGNIQEOFURP-UHFFFAOYSA-N
SMILES: O=C1OC2=CC3=C(C=CO3)C(OCCCCC3=CC=CC=C3)=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results