accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)N(R)HO(S)HO(R)

CAS RN 72741-87-8

IUPAC Name: (1S,2R,8R,8aR)-octahydroindolizine-1,2,8-triol
Synonyms: swainsonine (8a α)-isomer swainsonine (8 α,8a α)-isomer swainsonine swainsonine, (2 β,8a α)- swainsonine, (8 α,8a α)- (8 α,8a α)-swainsonine (8a α)-swainsonine (8 α)-isomer swainsonine swainsonine, (8 α)-isomer swainsonine, (8a α)-
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: FXUAIOOAOAVCGD-WCTZXXKLSA-N
SMILES: O[C@@H]1CN2CCC[C@@H](O)[C@@H]2[C@@H]1O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results