accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOO

CAS RN 72835-26-8

IUPAC Name: (2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl propanoate
Synonyms: (2,5-dioxopyrrol-1-yl)methyl propanoate
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: YXEWPGYLMHXLPS-UHFFFAOYSA-N
SMILES: CCC(=O)OCN1C(=O)C=CC1=O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results