accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNH3CCH3HN

CAS RN 73029-73-9

IUPAC Name: N-cyclohexyl-2-(3,5-dimethyl-1H-pyrazol-1-yl)-6-methylpyrimidin-4-amine
Synonyms: N-cyclohexyl-2-(3,5-dimethylpyrazol-1-yl)-6-methylpyrimidin-4-amine
Molecular Weight (g/mol): 285.4
Molecular Formula: C16H23N5
InChi Key: USEMRPYUFJNFQN-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=NC(C)=CC(NC2CCCCC2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results