accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73590-85-9

IUPAC Name: 6-methoxy-2-{[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulfanyl}-1H-1,3-benzodiazole
Synonyms: ufiprazole
Molecular Weight (g/mol): 329.42
Molecular Formula: C17H19N3O2S
InChi Key: XURCIPRUUASYLR-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(NC2=C1)SCC1=NC=C(C)C(OC)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results