accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 736991-21-2

IUPAC Name: 2-[(morpholin-4-yl)methyl]benzaldehyde
Synonyms: 2-(morpholin-4-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: YPZNTOVVELIJGI-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results