accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74-97-5

ClBr

CAS RN 74-97-5

IUPAC Name: bromo(chloro)methane
Synonyms: bromochloromethane fluorocarbon 1011 ccris 817 C-H2-br-cl methylene chlorobromide
Molecular Weight (g/mol): 129.38
Molecular Formula: CH2BrCl
InChi Key: JPOXNPPZZKNXOV-UHFFFAOYSA-N
SMILES: ClCBr
Molecular Weight (g/mol) 
  • (1)
  • (1)
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results