accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 7420-16-8

IUPAC Name: 3-(decyloxy)propanoic acid
Synonyms: 3-(decyloxy)propanoic acid
Molecular Weight (g/mol): 230.35
Molecular Formula: C13H26O3
InChi Key: ZYLHDTBQHXSLEZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results