accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(R)OPOOONCH3CH3H3COH3CO

CAS RN 74389-68-7

IUPAC Name: (2-{[(2R)-2-(acetyloxy)-3-(hexadecyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Synonyms: platelet activating factor platelet-activating factor C16-paf C16-paf acether
Molecular Weight (g/mol): 523.69
Molecular Formula: C26H54NO7P
InChi Key: HVAUUPRFYPCOCA-AREMUKBSSA-N
SMILES: CCCCCCCCCCCCCCCCOC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)