Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PAF (C16)
SDP

Catalog No. 29401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-401 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-401 Supplier Tocris Bioscience™ Supplier No. 2940/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Endogenous platelet-activating factor (PAF)

Endogenous platelet-activating factor (PAF) and ligand for PAF receptors. Produced by inflammatory cells and a potent chemoattractant for polymorphonuclear neutrophils. Induces increased vascular permeability.

Chemical Identifiers

CAS 74389-68-7
Molecular Formula C26H54NO7P

Specifications

Quantity 1 mg
Formula Weight 523.68
Purity >98%
Molecular Weight (g/mol) 523.69
Chemical Name or Material PAF (C16)
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.