CAS RN 7440-24-6
Strontium, distilled dendritic pieces, 99.95% (metals basis)
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 MDL Number: MFCD00134060 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N Synonym: strontium, elemental,unii-yzs2rpe8le,yzs2rpe8le,estroncio,granules, 19mm 0.76in & down metals basis,strontium, distilled dendritic pieces,atom,pieces,strontium, chunks,pieces, dendritic PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium granules, 20mm max, 99.0% (metals basis)
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 MDL Number: MFCD00134060 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N Synonym: strontium, elemental,unii-yzs2rpe8le,yzs2rpe8le,estroncio,granules, 19mm 0.76in & down metals basis,strontium, distilled dendritic pieces,atom,pieces,strontium, chunks,pieces, dendritic PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium granules, 19mm (0.76in) & down, 99% (metals basis)
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 MDL Number: MFCD00134060 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N Synonym: strontium, elemental,unii-yzs2rpe8le,yzs2rpe8le,estroncio,granules, 19mm 0.76in & down metals basis,strontium, distilled dendritic pieces,atom,pieces,strontium, chunks,pieces, dendritic PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium Standard For ICP, 1000μg/mL in 2% HCl, SPEX CertiPrep™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Sigma Aldrich Strontium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Agilent Technologies Strontium Standard, 1000μg/mL in water with dilute nitric acid, Ultra Scientific
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Sigma Aldrich Strontium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Strontium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Strontium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Agilent Technologies Strontium Standard, 10,000μg/mL water with dilute nitric acid, Ultra Scientific
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
MP Biomedicals, Inc Strontium, 99%, MP Biomedicals
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N Synonym: strontium, elemental,unii-yzs2rpe8le,yzs2rpe8le,estroncio,granules, 19mm 0.76in & down metals basis,strontium, distilled dendritic pieces,atom,pieces,strontium, chunks,pieces, dendritic PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]