CAS RN 74885-72-6
IUPAC Name:
3-(2-aminoethyl)-1H-indole-5-carboxamide; but-2-enedioic acid
Synonyms:
5-carboxamidotryptamine; butenedioic acid
Molecular Weight (g/mol):
319.32
Molecular Formula:
C15H17N3O5
InChi Key:
PZQZSWAOVAMBQM-UHFFFAOYSA-N
SMILES:
OC(=O)C=CC(O)=O.NCCC1=CNC2=CC=C(C=C12)C(N)=O
1
–
1
of
1
results
1
R&D Systems™ 5-Carboxamidotryptamine maleate
CAS: 74885-72-6 Molecular Formula: C11H13N3O·C4H4O4