accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7497-07-6

IUPAC Name: 3,4-dichloro-N-(pentan-2-yl)benzamide
Synonyms: 3,4-dichloro-N-(pentan-2-yl)benzamide
Molecular Weight (g/mol): 260.16
Molecular Formula: C12H15Cl2NO
InChi Key: ARDYECYBETXQFD-UHFFFAOYNA-N
SMILES: CCCC(C)NC(=O)C1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results