accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 75-07-0

IUPAC Name: acetaldehyde
Synonyms: acetaldehyde octowy aldehyd acetaldehyde (natural) acetaldehyde (substance) azetaldehyd aldehyde acetique aldeide acetica acetaldehydes acetic aldehyde rcra waste number U001
Molecular Weight (g/mol): 44.05
Molecular Formula: C2H4O
InChi Key: IKHGUXGNUITLKF-UHFFFAOYSA-N
SMILES: CC=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (10)
  • (6)
Quantity 
  • (5)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (10)
  • (2)
  • (1)
  • (6)
Physical Form 
  • (6)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (2)
  • (1)
  • (10)
  • (2)
  • (2)
  • (1)
  • (9)
  • (1)
  • (6)
  • (3)
  • (10)
  • (6)
  • (5)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (10)
  • (2)
  • (1)
  • (6)
  • (6)
  • (2)
  • (4)
  • (3)
  • (8)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)