Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dichloroethylene (stabilized with MEHQ) (in cylinder without valve), TCI America™
SDP

Catalog No. D0366250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0366250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0366250G Supplier TCI America Supplier No. D0366250G

missing translation for 'validMainframeMsg'

[To use this product charged in cylinder, a valve is required which is sold separately (Product Code:V0030)]

Chemical Identifiers

CAS 75-35-4
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.938
MDL Number MFCD00011653
InChI Key LGXVIGDEPROXKC-UHFFFAOYSA-N
Synonym vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene
PubChem CID 6366
ChEBI CHEBI:34031
IUPAC Name 1,1-dichloroethene
SMILES C=C(Cl)Cl

Specifications

Color Colorless
Boiling Point 31°C
UN Number 1303
Quantity 250 g
Formula Weight 96.94
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Dichloroethylene (stabilized with MEHQ) (in cylinder without valve) [To use this product charged
EINECSNumber (2)-0103
RTECSNumber KV9275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.