accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHNNNFFFNOCH3

CAS RN 752222-83-6

IUPAC Name: 3-ethyl-1-propyl-8-(1-{[3-(trifluoromethyl)phenyl]methyl}-1H-pyrazol-4-yl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 3-ethyl-1-propyl-8-(1-{[3-(trifluoromethyl)phenyl]methyl}pyrazol-4-yl)-7H-purine-2,6-dione
Molecular Weight (g/mol): 446.43
Molecular Formula: C21H21F3N6O2
InChi Key: KOYXXLLNCXWUNF-UHFFFAOYSA-N
SMILES: CCCN1C(=O)N(CC)C2=C(NC(=N2)C2=CN(CC3=CC(=CC=C3)C(F)(F)F)N=C2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results