accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNOSCl

CAS RN 75450-34-9

IUPAC Name: 7-chloro-5-(2-chlorophenyl)-1,2,3,5-tetrahydro-4,1-benzothiazepin-2-one
Synonyms: 7-chloro-5-(2-chlorophenyl)-3,5-dihydro-1H-4,1-benzothiazepin-2-one
Molecular Weight (g/mol): 324.22
Molecular Formula: C15H11Cl2NOS
InChi Key: KQEPIRKXSUIUTH-UHFFFAOYNA-N
SMILES: ClC1=CC=C2NC(=O)CSC(C3=CC=CC=C3Cl)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)