CAS RN 75859-03-9
IUPAC Name:
dihydrogen 9-{3-[(3R,5S)-3,5-dimethylpiperazin-1-yl]propyl}-9H-carbazole dichloride
Synonyms:
dihydrogen rimcazole dichloride
Molecular Weight (g/mol):
394.38
Molecular Formula:
C21H29Cl2N3
InChi Key:
ZWHRZGHGYMUSRM-VWDRLOGHSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].C[C@H]1CN(CCCN2C3=C(C=CC=C3)C3=C2C=CC=C3)C[C@@H](C)N1
1
–
1
of
1
results
1
R&D Systems™ Rimcazole dihydrochloride
CAS: 75859-03-9 Molecular Formula: C21H29Cl2N3