accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7598-26-7

CH3NHNH2NOO

CAS RN 7598-26-7

IUPAC Name: 2-amino-5-methyl-3-nitropyridin-1-ium
Synonyms: 2-amino-5-methyl-3-nitropyridin-1-ium
Molecular Weight (g/mol): 154.15
Molecular Formula: C6H8N3O2
InChi Key: ZJKGRSJMFDIRPX-UHFFFAOYSA-O
SMILES: CC1=C[NH+]=C(N)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results