accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2POHHOOOHO

CAS RN 76326-31-3

IUPAC Name: 2-amino-5-phosphonopentanoic acid
Synonyms: 2-amino-5-phosphovaleric acid apv acid app acid
Molecular Weight (g/mol): 197.13
Molecular Formula: C5H12NO5P
InChi Key: VOROEQBFPPIACJ-UHFFFAOYNA-N
SMILES: NC(CCCP(O)(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results