accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7643-08-5

CH3SSNH

CAS RN 7643-08-5

IUPAC Name: 2-(methylsulfanyl)-10H-phenothiazine
Synonyms: 2-(methylsulfanyl)-10H-phenothiazine
Molecular Weight (g/mol): 245.36
Molecular Formula: C13H11NS2
InChi Key: OBVKBOLDEFIQDP-UHFFFAOYSA-N
SMILES: CSC1=CC=C2SC3=CC=CC=C3NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results