accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77671-31-9

IUPAC Name: 4-methyl-5-[4-(methylsulfanyl)benzoyl]-2,3-dihydro-1H-imidazol-2-one
Synonyms: enoximone enoximonum fenoximone enoximona enoximone (product) perfan enoximone (substance)
Molecular Weight (g/mol): 248.3
Molecular Formula: C12H12N2O2S
InChi Key: ZJKNESGOIKRXQY-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)C(=O)C1=C(C)NC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results