Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Enoximone
GREENER_CHOICESDP

Catalog No. 169350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-935-0 50 mg
16-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-935-0 Supplier Tocris Bioscience™ Supplier No. 1693/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

PDE3 inhibitor

Inhibitor of type III phosphodiesterase (PDE3). Increases intracellular cyclic AMP (cAMP) concentrations and enhances myocardial contractility. Also induces concentration-dependent vasodilation in vitro.

Chemical Identifiers

CAS 77671-31-9
Molecular Formula C12H12N2O2S

Specifications

Quantity 50 mg
Formula Weight 248.3
Purity >99%
Molecular Weight (g/mol) 248.30
Chemical Name or Material Enoximone
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.