accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClBi

CAS RN 7787-60-2

IUPAC Name: bismuth(3+) trichloride
Synonyms: bismuth chloride
Molecular Weight (g/mol): 315.33
Molecular Formula: BiCl3
InChi Key: JHXKRIRFYBPWGE-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Bi+3]
Quantity 
  • (4)
  • (1)
  • (10)
  • (1)
  • (5)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (15)
  • (9)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
Grade 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (4)
  • (5)
  • (2)
  • (14)
  • (4)
  • (1)
  • (10)
  • (1)
  • (5)
  • (1)
  • (3)
  • (15)
  • (9)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
  • (5)
  • (2)
  • (2)