accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHONHN

CAS RN 78538-74-6

IUPAC Name: N-methyl-9H-pyrido[3,4-b]indole-3-carboxamide
Synonyms: N-methyl-9H-pyrido[3,4-b]indole-3-carboxamide
Molecular Weight (g/mol): 225.25
Molecular Formula: C13H11N3O
InChi Key: QMCOPDWHWYSJSA-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC2=C(NC3=C2C=CC=C3)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)