CAS RN 78860-34-1
IUPAC Name:
2,5-dihydroxy-3-[7-(3-methylbut-2-en-1-yl)-1H-indol-3-yl]-6-[2-(2-methylbut-3-en-2-yl)-1H-indol-3-yl]cyclohexa-2,5-diene-1,4-dione
Synonyms:
2,5-dihydroxy-3-[7-(3-methylbut-2-en-1-yl)-1H-indol-3-yl]-6-[2-(2-methylbut-3-en-2-yl)-1H-indol-3-yl]cyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol):
506.6
Molecular Formula:
C32H30N2O4
InChi Key:
XMGNJVXBPZAETK-UHFFFAOYSA-N
SMILES:
CC(C)=CCC1=C2NC=C(C2=CC=C1)C1=C(O)C(=O)C(C2=C(NC3=CC=CC=C23)C(C)(C)C=C)=C(O)C1=O
1
–
1
of
1
results
1
R&D Systems™ Demethylasterriquinone B1
CAS: 78860-34-1 Molecular Formula: C32H30N2O4