accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79-04-9

IUPAC Name: 2-chloroacetyl chloride
Synonyms: chloroacetyl chloride chlorid kyseliny chloroctove chlorure de chloracetyle
Molecular Weight (g/mol): 112.94
Molecular Formula: C2H2Cl2O
InChi Key: VGCXGMAHQTYDJK-UHFFFAOYSA-N
SMILES: ClCC(Cl)=O
Molecular Weight (g/mol) 
  • (1)
  • (10)
Percent Purity 
  • (3)
  • (8)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (5)
  • (1)
  • (10)
  • (3)
  • (8)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)