CAS RN 79183-19-0
IUPAC Name:
1-[(3,4-dichlorophenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione
Synonyms:
1-[(3,4-dichlorophenyl)methyl]indole-2,3-dione
Molecular Weight (g/mol):
306.14
Molecular Formula:
C15H9Cl2NO2
InChi Key:
KGRJPLRFGLMQMV-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(CN2C(=O)C(=O)C3=CC=CC=C23)C=C1Cl
1
–
2
of
2
results
1
Apoptosis Activator 2, Thermo Scientific Chemicals
CAS: 79183-19-0 Molecular Formula: C15H9Cl2NO2,C15H9Cl2NO2 Synonym: 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione
2
Tocris Bioscience™ Apoptosis Activator 2
CAS: 79183-19-0 Molecular Formula: C15H9Cl2NO2