accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79893-89-3

OHOOONH(S)(R)CH3H3C

CAS RN 79893-89-3

IUPAC Name: methyl (2S,3R)-3-hydroxy-2-(phenylformamido)butanoate
Synonyms: methyl (2S,3R)-3-hydroxy-2-(phenylformamido)butanoate
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: KHOWDUMYRBCHAC-SCZZXKLOSA-N
SMILES: COC(=O)[C@@H](NC(=O)C1=CC=CC=C1)[C@@H](C)O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)