accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80-00-2

IUPAC Name: 1-(benzenesulfonyl)-4-chlorobenzene
Synonyms: sulphenone sulfenone
Molecular Weight (g/mol): 252.71
Molecular Formula: C12H9ClO2S
InChi Key: OFCFYWOKHPOXKF-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results