Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Stock Standard Mixture (2 components), Restek™
SDP

Catalog No. 06802062 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-062 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-062 Supplier Restek Corporation Supplier No. 31630
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Compounds:

  • Diesel #2 (additive-free) (68334-30-5)
  • Mineral oil (additive-free [i.e., USP grade] or
  • C18–C32 (retention time range) (8042-47-5)
Reference mixtures for ISO/DIS 9377 (German H-53), a gas chromatography, flame ionization detection (GC-FID) method.
For GC analysis of total petroleum hydrocarbons (TPH) in water

Chemical Identifiers

CAS 8012-95-1 , 110-82-7 , 68334-30-5
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *

Specifications

CAS Max % 99.00000
CAS Min % 0.5
Ignition Point 245°C
Odor Pungent
Vapor Density 2.9 (Air = 1)
Flash Point −4°C
Specific Gravity 0.779 to 0.784 g/cm3 at 20°C
Concentration 5,000 μg/mL each in cyclohexane, 1 mL/ampul (prepares 8 mL of 1.25 μg/μL calibration curve high point). Total hydrocarbon concentration is 10,000 μg/mL.
Color Colorless
Melting Point 6.5°C
Type Multi-component Standard
Quantity 1 mL
Show More Show Less
Hazard Category Danger
Hazard Statement Keep away from heat/sparks/open flames/hot surfaces.
No smoking. Ground/bond container and receiving equipment
Use explosion - proof electrical/ventilation and lighting equipment
Use only non - sparking tools
Take precautionary measures against static discharge
Avoid breathing dust/fume/gas/mist/vapours/spray
Wash hands and skin thoroughly after handling
Use only outdoors or in a well - ventilated area
Avoi d release to the environment
Wear protective gloves/protective clothing/eye protection/face protection
ShelfLife 6 months
Recommended Storage Store in a well ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.