accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOHOHOHONNNH2ClH2O

CAS RN 81012-94-4

IUPAC Name: 2-(6-amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol hydrate
Synonyms: 2-chloroadenosine hydrate
Molecular Weight (g/mol): 319.7
Molecular Formula: C10H14ClN5O5
InChi Key: CSVJEVDOHIWPDO-UHFFFAOYNA-N
SMILES: O.NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC(Cl)=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results