accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOFHNOClH2NOCH3

CAS RN 81098-60-4

IUPAC Name: 4-amino-5-chloro-N-{1-[3-(4-fluorophenoxy)propyl]-3-methoxypiperidin-4-yl}-2-methoxybenzamide
Synonyms: cisapride cisawal presid guptro propulsin propulsid kaudalit C23H29ClFN3O4 cipril acenalin
Molecular Weight (g/mol): 465.95
Molecular Formula: C23H29ClFN3O4
InChi Key: DCSUBABJRXZOMT-UHFFFAOYNA-N
SMILES: COC1CN(CCCOC2=CC=C(F)C=C2)CCC1NC(=O)C1=CC(Cl)=C(N)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)