accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNHNCl

CAS RN 81226-60-0

IUPAC Name: 4-(4-chlorophenyl)-1-[(1H-indol-3-yl)methyl]piperidin-4-ol
Synonyms: 4-(4-chlorophenyl)-1-(1H-indol-3-ylmethyl)piperidin-4-ol
Molecular Weight (g/mol): 340.85
Molecular Formula: C20H21ClN2O
InChi Key: LLBLNMUONVVVPG-UHFFFAOYSA-N
SMILES: OC1(CCN(CC2=CNC3=CC=CC=C23)CC1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)