accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNONNCH3H3CO(R)(R)(R)HNNH2C

CAS RN 81409-90-7

IUPAC Name: 1-[3-(dimethylamino)propyl]-3-ethyl-1-[(2R,4R,7R)-6-(prop-2-en-1-yl)-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carbonyl]urea
Synonyms: cabergoline cabergolinum cabaser cabergolina dostinex 卡麦角林
Molecular Weight (g/mol): 451.62
Molecular Formula: C26H37N5O2
InChi Key: KORNTPPJEAJQIU-KJXAQDMKSA-N
SMILES: CCNC(=O)N(CCCN(C)C)C(=O)[C@@H]1C[C@H]2[C@@H](CC3=CNC4=CC=CC2=C34)N(CC=C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)