Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cabergoline
GREENER_CHOICESDP

Catalog No. 266450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-645-0 50 mg
26-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-645-0 Supplier R&D Systems™ Supplier No. 2664/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective D2-like agonist

Cabergoline is a selective D2-like dopamine receptor agonist (Ki values are 0.7, 1.5, 9.0 and 165 nM for D2, D3, D4 and D5 receptors respectively) that also displays high affinity for several serotonin receptor subtypes (Ki = 1.2 - 20.0 nM for 5-HT1A, 5-HT1D, 5-HT2A and 5-HT2B). Inhibits secretion of prolactin and growth hormone and reverses levodopa -induced dyskinesias in Parkinsonian monkeys.

Chemical Identifiers

CAS 81409-90-7
Molecular Formula C26H37N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 451.6
Purity >99%
Molecular Weight (g/mol) 451.615
Chemical Name or Material Cabergoline
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.