accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3OSOOFFFSiCH3H3CCH3OSOOFFF

CAS RN 828282-80-0

IUPAC Name: 4'-(trifluoromethanesulfonyloxy)-3,3'-bis(trimethylsilyl)-[1,1'-biphenyl]-4-yl trifluoromethanesulfonate
Synonyms: 4'-(trifluoromethanesulfonyloxy)-3,3'-bis(trimethylsilyl)-[1,1'-biphenyl]-4-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 594.69
Molecular Formula: C20H24F6O6S2Si2
InChi Key: FDAGZCNNWNNCEK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=C(OS(=O)(=O)C(F)(F)F)C=CC(=C1)C1=CC(=C(OS(=O)(=O)C(F)(F)F)C=C1)[Si](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results