accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82938-50-9

CH3H3CCH3SiH3CCH3NOOOH

CAS RN 82938-50-9

IUPAC Name: 1-(tert-butyldimethylsilyl)-4-oxoazetidine-2-carboxylic acid
Synonyms: 1-(tert-butyldimethylsilyl)-4-oxoazetidine-2-carboxylic acid
Molecular Weight (g/mol): 229.35
Molecular Formula: C10H19NO3Si
InChi Key: LIEWITJXZYCDLE-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)N1C(CC1=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results